accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 103365-47-5

IUPAC Name: (3S)-3-{[(tert-butoxy)carbonyl]amino}-3-phenylpropanoate
Synonyms: (3S)-3-[(tert-butoxycarbonyl)amino]-3-phenylpropanoate
Molecular Weight (g/mol): 264.3
Molecular Formula: C14H18NO4
InChi Key: JTNQFJPZRTURSI-NSHDSACASA-M
SMILES: CC(C)(C)OC(=O)N[C@@H](CC([O-])=O)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results