accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

TiTiOSOOOOSOOOOSOOO

CAS RN 10343-61-0

IUPAC Name: dititanium(3+) trisulfate
Synonyms: dititanium(3+) trisulfate
Molecular Weight (g/mol): 383.9
Molecular Formula: O12S3Ti2
InChi Key: SOBXOQKKUVQETK-UHFFFAOYSA-H
SMILES: [Ti+3].[Ti+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
pH 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results