accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 10344-94-2

IUPAC Name: (2S,3S,4S,5R,6S)-3,4,5-trihydroxy-6-(4-nitrophenoxy)oxane-2-carboxylate
Synonyms: (2S,3S,4S,5R,6S)-3,4,5-trihydroxy-6-(4-nitrophenoxy)oxane-2-carboxylate
Molecular Weight (g/mol): 314.23
Molecular Formula: C12H12NO9
InChi Key: QSUILVWOWLUOEU-GOVZDWNOSA-M
SMILES: O[C@@H]1[C@@H](O)[C@H](OC2=CC=C(C=C2)[N+]([O-])=O)O[C@@H]([C@H]1O)C([O-])=O
Molecular Weight (g/mol) 
  • (4)
  • (1)
Physical Form 
  • (3)
  • (2)
Quantity 
  • (3)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results