accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1034617-86-1

CH3NNOCH3NOO

CAS RN 1034617-86-1

IUPAC Name: 1-(4-methoxy-3-nitrophenyl)-4-methylpiperazine
Synonyms: 1-(4-methoxy-3-nitrophenyl)-4-methylpiperazine
Molecular Weight (g/mol): 251.29
Molecular Formula: C12H17N3O3
InChi Key: XHYHNDLFYCMLQK-UHFFFAOYSA-N
SMILES: COC1=C(C=C(C=C1)N1CCN(C)CC1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results