accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HO(R)OONOO(S)OH(S)HO(S)HO

CAS RN 10357-27-4

IUPAC Name: (2R,3S,4S,5S)-2-(hydroxymethyl)-6-(4-nitrophenoxy)oxane-3,4,5-triol
Synonyms: (2R,3S,4S,5S)-2-(hydroxymethyl)-6-(4-nitrophenoxy)oxane-3,4,5-triol
Molecular Weight (g/mol): 301.25
Molecular Formula: C12H15NO8
InChi Key: IFBHRQDFSNCLOZ-IKQSSVLVSA-N
SMILES: OC[C@H]1OC(OC2=CC=C(C=C2)[N+]([O-])=O)[C@@H](O)[C@@H](O)[C@@H]1O
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)