accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3SOHNNNOFFF

CAS RN 103577-45-3

IUPAC Name: 2-{[3-methyl-4-(2,2,2-trifluoroethoxy)pyridin-2-yl]methanesulfinyl}-1H-1,3-benzodiazole
Synonyms: lansoprazole Lanz prevpac monolitum lansophed wyeth brand of lansoprazole tecnobio brand of lansoprazole prezal prosogan ilsatec
Molecular Weight (g/mol): 369.36
Molecular Formula: C16H14F3N3O2S
InChi Key: MJIHNNLFOKEZEW-UHFFFAOYNA-N
SMILES: CC1=C(CS(=O)C2=NC3=CC=CC=C3N2)N=CC=C1OCC(F)(F)F
Molecular Weight (g/mol) 
  • (5)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (3)
Certifications/Compliance 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (3)
  • (5)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)