accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 10361-46-3

ONOOBiO

CAS RN 10361-46-3

IUPAC Name: (nitrooxy)bismuthanone
Synonyms: (nitrooxy)bismuthanone
Molecular Weight (g/mol): 286.98
Molecular Formula: BiNO4
InChi Key: OGCYGKRWXWQOAK-UHFFFAOYSA-N
SMILES: [O-][N+](=O)O[Bi]=O
Grade 
  • (3)
Percent Purity 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)