accessibility menu, dialog, popup

UserName

CAS RN 10361-91-8

CAS RN 10361-91-8

IUPAC Name: ytterbium(3+) trichloride
Synonyms: ytterbium(III) chloride
Molecular Weight (g/mol): 279.4
Molecular Formula: Cl3Yb
InChi Key: CKLHRQNQYIJFFX-UHFFFAOYSA-K
SMILES: [Cl-].[Cl-].[Cl-].[Yb+3]
Physical Form 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)