accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3PH3CCH3

CAS RN 1038-95-5

IUPAC Name: tris(4-methylphenyl)phosphane
Synonyms: tris(4-methylphenyl)phosphane
Molecular Weight (g/mol): 304.37
Molecular Formula: C21H21P
InChi Key: WXAZIUYTQHYBFW-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)P(C1=CC=C(C)C=C1)C1=CC=C(C)C=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results