accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ONONO

CAS RN 10389-51-2

IUPAC Name: 4-(4-nitrophenyl)morpholine
Synonyms: morpholine, 4-(4-nitrophenyl)-
Molecular Weight (g/mol): 208.22
Molecular Formula: C10H12N2O3
InChi Key: IAJDSUYFELYZCS-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(C=C1)N1CCOCC1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results