accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 103956-09-8

IUPAC Name: 3,4-diaminobenzohydrazide
Synonyms: 3,4-diaminobenzohydrazide
Molecular Weight (g/mol): 166.18
Molecular Formula: C7H10N4O
InChi Key: YGCXQTYRSKMILM-UHFFFAOYSA-N
SMILES: NNC(=O)C1=CC(N)=C(N)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results