accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

Cl

CAS RN 104-52-9

IUPAC Name: (3-chloropropyl)benzene
Synonyms: 3-phenyl-1-chloropropane
Molecular Weight (g/mol): 154.64
Molecular Formula: C9H11Cl
InChi Key: XZBXAYCCBFTQHH-UHFFFAOYSA-N
SMILES: ClCCCC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (1)
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results