CAS RN 104-61-0
IUPAC Name:
(5S)-5-pentyloxolan-2-one
Synonyms:
gamma-nonalactone fema number 2781 coconut aldehyde aldehyde C-18 1,4-nonalolide 4-nonanolide gamma-nonyllactone 4-pentyl-butanolide gamma-amyl-gamma-butyrolactone nonan-4-olide
Molecular Weight (g/mol):
156.23
Molecular Formula:
C9H16O2
InChi Key:
OALYTRUKMRCXNH-QMMMGPOBSA-N
SMILES:
CCCCC[C@H]1CCC(=O)O1
1
–
1
of
1
results
1
rac-gamma-Nonanolactone, TRC
CAS: 104-61-0 Molecular Formula: C9 H16 O2 Molecular Weight (g/mol): 156.22 Synonym: 4-Nonanolide,4-Pentyl-butanolide,γ-Amylbutyrolactone,γ-Nonanolide,Aldehyde C-18,Apricolin,Cocos aldehyde,Dihydro-5-pentyl-2(3H)-furanone,Prunolide IUPAC Name: 5-pentyloxolan-2-one SMILES: CCCCCC1CCC(=O)O1