accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NaCH3ONOHN(R)(R)SNOOOSOOSH2NN(Z)

CAS RN 104010-37-9

IUPAC Name: sodium (6R,7R)-7-[(2Z)-2-(2-amino-1,3-thiazol-4-yl)-2-(methoxyimino)acetamido]-3-[(furan-2-carbonylsulfanyl)methyl]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
Synonyms: ceftiofur
Molecular Weight (g/mol): 545.53
Molecular Formula: C19H16N5NaO7S3
InChi Key: RFLHUYUQCKHUKS-JUODUXDSSA-M
SMILES: [Na+].CO\N=C(/C(=O)N[C@H]1[C@H]2SCC(CSC(=O)C3=CC=CO3)=C(N2C1=O)C([O-])=O)C1=CSC(N)=N1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Color 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results