accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1040683-08-6

IUPAC Name: ethyl 1-(trifluoromethyl)cyclobutane-1-carboxylate
Synonyms: ethyl 1-(trifluoromethyl)cyclobutane-1-carboxylate
Molecular Weight (g/mol): 196.17
Molecular Formula: C8H11F3O2
InChi Key: AYUISFJYNKIHDD-UHFFFAOYSA-N
SMILES: CCOC(=O)C1(CCC1)C(F)(F)F
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results