accessibility menu, dialog, popup

UserName

CAS RN 104163-39-5

CAS RN 104163-39-5

IUPAC Name: 1-(4-methylthiophen-2-yl)methanamine
Synonyms: 1-(4-methylthiophen-2-yl)methanamine
Molecular Weight (g/mol): 127.21
Molecular Formula: C6H9NS
InChi Key: CKQHNKAVFNDGMK-UHFFFAOYSA-N
SMILES: CC1=CSC(CN)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results