CAS RN 10421-48-4
CAS RN 10421-48-4
IUPAC Name:
iron(3+) tris(nitric acid) nonahydrate
Synonyms:
ferric nitrate
Molecular Weight (g/mol):
407.01
Molecular Formula:
FeH21N3O18
InChi Key:
MUSSTCMSWRZDLN-UHFFFAOYSA-N
SMILES:
O.O.O.O.O.O.O.O.O.[Fe+3].O[N+]([O-])=O.O[N+]([O-])=O.O[N+]([O-])=O
1
–
3
of
3
results
1
Iron Standard Metal Solution 1000ppm, for AAS, Solution HNO3 1M, Fisher Chemical™
CAS: 10421-48-4 Molecular Formula: FeN3O9 Molecular Weight (g/mol): 241.857 MDL Number: 11003 InChI Key: VCJMYUPGQJHHFU-UHFFFAOYSA-N Synonym: ferric nitrate,iron trinitrate,nitric acid, iron 3+ salt,unii-n8h8402xob,iron nitrate fe no3 3,iron iii nitrate, anhydrous,hsdb 451,iron 3+ trinitrate,nitric acid, iron 3+ salt 3:1 PubChem CID: 25251 IUPAC Name: iron(3+);trinitrate SMILES: [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[Fe+3]
3
Honeywell Fluka Iron AAS Standard, Fluka™
Certified Reference Material, 1000 mg/L Fe in HNO3 2%