accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OOOHHO(E/Z)

CAS RN 104236-78-4

IUPAC Name: 1-(2,4-dihydroxyphenyl)-3-(2-methoxyphenyl)prop-2-en-1-one
Synonyms: 1-(2,4-dihydroxyphenyl)-3-(2-methoxyphenyl)prop-2-en-1-one
Molecular Weight (g/mol): 270.28
Molecular Formula: C16H14O4
InChi Key: ODLVGCCGMXGMGZ-UHFFFAOYSA-N
SMILES: COC1=CC=CC=C1C=CC(=O)C1=C(O)C=C(O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results