accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1044256-04-3

IUPAC Name: 1-[2-(1,3-dioxolan-2-yl)phenyl]-2,5-dimethylphospholane
Synonyms: 1-[2-(1,3-dioxolan-2-yl)phenyl]-2,5-dimethylphospholane
Molecular Weight (g/mol): 264.31
Molecular Formula: C15H21O2P
InChi Key: JAKXTLMNIHPLEU-UHFFFAOYNA-N
SMILES: CC1CCC(C)P1C1=CC=CC=C1C1OCCO1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results