accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 10458-14-7

H3CCH3CH3O

CAS RN 10458-14-7

IUPAC Name: 5-methyl-2-(propan-2-yl)cyclohexan-1-one
Synonyms: menthone isomenthone
Molecular Weight (g/mol): 154.25
Molecular Formula: C10H18O
InChi Key: NFLGAXVYCFJBMK-UHFFFAOYNA-N
SMILES: CC(C)C1CCC(C)CC1=O
Boiling Point 
  • (3)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)