accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1046-56-6

IUPAC Name: 5,6-diphenyl-3-(pyridin-2-yl)-1,2,4-triazine
Synonyms: 5,6-diphenyl-3-(pyridin-2-yl)-1,2,4-triazine
Molecular Weight (g/mol): 310.36
Molecular Formula: C20H14N4
InChi Key: OTMYLOBWDNFTLO-UHFFFAOYSA-N
SMILES: C1=CC=C(C=C1)C1=NN=C(N=C1C1=CC=CC=C1)C1=CC=CC=N1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results