CAS RN 104663-56-1
IUPAC Name:
manganese(3+) hydrate phosphate
Synonyms:
manganese(3+) hydrate phosphate
Molecular Weight (g/mol):
167.92
Molecular Formula:
H2MnO5P
InChi Key:
VRHPRPVJMYLUBR-UHFFFAOYSA-K
SMILES:
O.[Mn+3].[O-]P([O-])([O-])=O
1
–
1
of
1
results
1
Manganese(III) phosphate hydrate, 99%
CAS: 104663-56-1 Molecular Formula: MnO4P Molecular Weight (g/mol): 149.91 MDL Number: MFCD00209658 InChI Key: BECVLEVEVXAFSH-UHFFFAOYSA-K PubChem CID: 71317435 SMILES: [Mn+3].[O-]P([O-])([O-])=O