CAS RN 104770-18-5
IUPAC Name:
ethyl 3-aminobicyclo[2.2.1]hept-5-ene-2-carboxylate hydrochloride
Synonyms:
ethyl 3-aminobicyclo[2.2.1]hept-5-ene-2-carboxylate hydrochloride
Molecular Weight (g/mol):
217.69
Molecular Formula:
C10H16ClNO2
InChi Key:
PBLUKJJZYDTALO-UHFFFAOYNA-N
SMILES:
Cl.CCOC(=O)C1C(N)C2CC1C=C2
1
–
1
of
1
results
1
Ethyl 3-exo-aminobicyclo[2.2.1]hept-5-ene-2-exo-carboxylate hydrochloride, 99%, Thermo Scientific™
CAS: 104770-18-5 Molecular Formula: C10H16ClNO2 Molecular Weight (g/mol): 217.69 MDL Number: MFCD00144272 InChI Key: PBLUKJJZYDTALO-UHFFFAOYNA-N Synonym: ethyl 3-exo-aminobicyclo 2.2.1 hept-5-ene-2-carboxylate hydrochloride,ethyl 3-exo-aminobicyclo 2.2.1 hept-5-ene-2-exo-carboxylate hydrochloride PubChem CID: 71528337 IUPAC Name: ethyl (1S,2S,3R,4R)-3-aminobicyclo[2.2.1]hept-5-ene-2-carboxylate;hydrochloride SMILES: Cl.CCOC(=O)C1C(N)C2CC1C=C2