accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 10485-09-3

IUPAC Name: 2-bromo-1H-indene
Synonyms: 2-bromo-1H-indene
Molecular Weight (g/mol): 195.06
Molecular Formula: C9H7Br
InChi Key: CCUYEVNCRQDQRF-UHFFFAOYSA-N
SMILES: BrC1=CC2=CC=CC=C2C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results