accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 10486-61-0

IUPAC Name: 3-iodothiophene
Synonyms: 3-iodothiophene
Molecular Weight (g/mol): 210.03
Molecular Formula: C4H3IS
InChi Key: WGKRMQIQXMJVFZ-UHFFFAOYSA-N
SMILES: IC1=CSC=C1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results