accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HNNNSOOOHSOOOSOOOHOONSOOOSOOOSOOOHONaNaNaNaNaNa(E/Z)(E/Z)(E/Z)

CAS RN 104869-31-0

IUPAC Name: hexasodium 3-[({3-[(4,8-disulfo-6-sulfonatonaphthalen-1-yl)carboximidato]phenyl}carboximidato)amino]-N-(8-sulfo-4,6-disulfonatonaphthalen-1-yl)benzene-1-carboximidate
Synonyms: hexasodium 3-[({3-[(4,8-disulfo-6-sulfonatonaphthalen-1-yl)carboximidato]phenyl}carboximidato)amino]-N-(8-sulfo-4,6-disulfonatonaphthalen-1-yl)benzenecarboximidate
Molecular Weight (g/mol): 1162.85
Molecular Formula: C35H20N4Na6O21S6
InChi Key: FMQURVHYTBGYSQ-UHFFFAOYSA-H
SMILES: [Na+].[Na+].[Na+].[Na+].[Na+].[Na+].OS(=O)(=O)C1=CC=C(N=C([O-])C2=CC(=CC=C2)N=C([O-])NC2=CC=CC(=C2)C([O-])=NC2=CC=C(C3=CC(=CC(=C23)S(O)(=O)=O)S([O-])(=O)=O)S([O-])(=O)=O)C2=C(C=C(C=C12)S([O-])(=O)=O)S(O)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results