accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OPSOONaNaNa

CAS RN 10489-48-2

IUPAC Name: trisodium hydrate phosphorothioate
Synonyms: trisodium hydrate thiophosphate
Molecular Weight (g/mol): 198.02
Molecular Formula: H2Na3O4PS
InChi Key: VYYYFZBRAKXIPU-UHFFFAOYSA-K
SMILES: O.[Na+].[Na+].[Na+].[O-]P([O-])([O-])=S
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
Grade 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)