CAS RN 104987-11-3
IUPAC Name:
(1R,9S,12S,13R,14S,17R,18E,21S,23S,24R,25S,27R)-1,14-dihydroxy-12-[(1E)-1-[(3R,4R)-4-hydroxy-3-methoxycyclohexyl]prop-1-en-2-yl]-23,25-dimethoxy-13,19,21,27-tetramethyl-17-(prop-2-en-1-yl)-11,28-dioxa-4-azatricyclo[22.3.1.0⁴,⁹]octacos-18-ene-2,3,10,16-tetrone hydrate
Synonyms:
tacrolimus
Molecular Weight (g/mol):
822.05
Molecular Formula:
C44H71NO13
InChi Key:
NWJQLQGQZSIBAF-HZXKVGIDSA-N
SMILES:
O.CO[C@@H]1CC(CC[C@H]1O)\C=C(/C)[C@H]1OC(=O)[C@@H]2CCCCN2C(=O)C(=O)[C@]2(O)O[C@@H]([C@H](C[C@H]2C)OC)[C@H](C[C@@H](C)C\C(C)=C\[C@@H](CC=C)C(=O)C[C@H](O)[C@H]1C)OC
1
–
3
of
3
results
1
Thermo Scientific Chemicals Tacrolimus, 99+%
CAS: 104987-11-3 Molecular Formula: C44H69NO12 Synonym: FK-506; Fugimycin
2
Tacrolimus (FK506), TRC
CAS: 104987-11-3 Molecular Formula: C44 H69 N O12 Molecular Weight (g/mol): 804.02 SMILES: CO[C@@H]1C[C@@H](CC[C@H]1O)\C=C(/C)\[C@H]2OC(=O)[C@@H]3CCCCN3C(=O)C(=O)[C@]4(O)O[C@H]([C@H](C[C@@H](C)C\C(=C\[C@@H](CC=C)C(=O)C[C@H](O)[C@H]2C)\C)OC)[C@H](C[C@H]4C)OC
3