accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3C(R)CH3(S)H3C(S)OCH3(R)O(R)HO(R)H3C(S)OCH3OON(S)OO(S)(R)CH3(S)HOOCH3(R)(R)HO(R)OCH3(E)(Z)

CAS RN 104987-12-4

IUPAC Name: (1R,9S,12S,13R,14S,17R,18E,21S,23S,24R,25S,27R)-17-ethyl-1,14-dihydroxy-12-[(1E)-1-[(1R,3R,4R)-4-hydroxy-3-methoxycyclohexyl]prop-1-en-2-yl]-23,25-dimethoxy-13,19,21,27-tetramethyl-11,28-dioxa-4-azatricyclo[22.3.1.0⁴,⁹]octacos-18-ene-2,3,10,16-tetrone
Synonyms: immunomycin
Molecular Weight (g/mol): 792.02
Molecular Formula: C43H69NO12
InChi Key: ZDQSOHOQTUFQEM-NURRSENYSA-N
SMILES: CC[C@@H]1\C=C(C)\C[C@H](C)C[C@H](OC)[C@H]2O[C@](O)([C@H](C)C[C@@H]2OC)C(=O)C(=O)N2CCCC[C@H]2C(=O)O[C@@H]([C@H](C)[C@@H](O)CC1=O)C(\C)=C\[C@@H]1CC[C@@H](O)[C@@H](C1)OC
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)