accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ON

CAS RN 105-07-7

IUPAC Name: 4-formylbenzonitrile
Synonyms: benzonitrile, 4-formyl-
Molecular Weight (g/mol): 131.13
Molecular Formula: C8H5NO
InChi Key: WZWIQYMTQZCSKI-UHFFFAOYSA-N
SMILES: O=CC1=CC=C(C=C1)C#N
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)
Percent Purity 
  • (1)
  • (3)
  • (2)
Boiling Point 
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)
  • (1)
  • (3)
  • (2)
  • (3)
  • (3)