accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 105-09-9

IUPAC Name: [4-(sulfanylmethyl)phenyl]methanethiol
Synonyms: [4-(sulfanylmethyl)phenyl]methanethiol
Molecular Weight (g/mol): 170.29
Molecular Formula: C8H10S2
InChi Key: IYPNRTQAOXLCQW-UHFFFAOYSA-N
SMILES: SCC1=CC=C(CS)C=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results