accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 105-46-4

IUPAC Name: butan-2-yl acetate
Synonyms: sec-butyl acetate S-butylacetate
Molecular Weight (g/mol): 116.16
Molecular Formula: C6H12O2
InChi Key: DCKVNWZUADLDEH-UHFFFAOYNA-N
SMILES: CCC(C)OC(C)=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results