accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 105-55-5

IUPAC Name: 1,3-diethylthiourea
Synonyms: N,N-diethylthiourea
Molecular Weight (g/mol): 132.23
Molecular Formula: C5H12N2S
InChi Key: FLVIGYVXZHLUHP-UHFFFAOYSA-N
SMILES: CCNC(=S)NCC
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results