CAS RN 105-59-9
CAS RN 105-59-9
IUPAC Name:
2-[(2-hydroxyethyl)(methyl)amino]ethan-1-ol
Synonyms:
N-methyldiethanolamine
Molecular Weight (g/mol):
119.16
Molecular Formula:
C5H13NO2
InChi Key:
CRVGTESFCCXCTH-UHFFFAOYSA-N
SMILES:
CN(CCO)CCO
1
–
1
of
1
results
1
Thermo Scientific Acros N-Methyldiethanolamine, 99+%, Thermo Scientific Chemicals
CAS: 105-59-9 Molecular Formula: C5H13NO2 Molecular Weight (g/mol): 119.16 MDL Number: MFCD00002848 InChI Key: CRVGTESFCCXCTH-UHFFFAOYSA-N Synonym: n-methyldiethanolamine,methyldiethanolamine,2,2'-methylimino diethanol,ethanol, 2,2'-methylimino bis,methyl diethanolamine,n-methyliminodiethanol,n-methyl-2,2'-iminodiethanol,n-methylaminodiglycol,2,2'-methyliminodiethanol,n-methyldiethanolimine PubChem CID: 7767 SMILES: CN(CCO)CCO