accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COOCH3CH3

CAS RN 105-79-3

IUPAC Name: 2-methylpropyl hexanoate
Synonyms: isobutyl hexanoate
Molecular Weight (g/mol): 172.27
Molecular Formula: C10H20O2
InChi Key: UXUPPWPIGVTVQI-UHFFFAOYSA-N
SMILES: CCCCCC(=O)OCC(C)C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results