accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OOH3CH3CH3C(E)

CAS RN 105-87-3

IUPAC Name: (2E)-3,7-dimethylocta-2,6-dien-1-yl acetate
Synonyms: geranyl acetate fema number 2509 geranyl acetate A citryl acetate geranyl acetate (natural) ccris 877 bay pine (oyster) oil
Molecular Weight (g/mol): 196.29
Molecular Formula: C12H20O2
InChi Key: HIGQPQRQIQDZMP-DHZHZOJOSA-N
SMILES: CC(C)=CCC\C(C)=C\COC(C)=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)