accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OOOO

CAS RN 105-95-3

IUPAC Name: 1,4-dioxacycloheptadecane-5,17-dione
Synonyms: ethylene brassylate
Molecular Weight (g/mol): 270.37
Molecular Formula: C15H26O4
InChi Key: XRHCAGNSDHCHFJ-UHFFFAOYSA-N
SMILES: O=C1CCCCCCCCCCCC(=O)OCCO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results