accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COHNOH3C

CAS RN 10514-70-2

IUPAC Name: 2,2-dimethyl-3,4-dihydro-2H-1,4-benzoxazin-3-one
Synonyms: 2,2-dimethyl-4H-1,4-benzoxazin-3-one
Molecular Weight (g/mol): 177.2
Molecular Formula: C10H11NO2
InChi Key: LJRUKUQEIHJTEL-UHFFFAOYSA-N
SMILES: CC1(C)OC2=CC=CC=C2NC1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results