accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 10523-68-9

IUPAC Name: hydrogen adamantan-2-amine chloride
Synonyms: hydrogen adamantan-2-amine chloride
Molecular Weight (g/mol): 187.71
Molecular Formula: C10H18ClN
InChi Key: WLDWDRZITJEWRJ-UHFFFAOYSA-N
SMILES: [H+].[Cl-].NC1C2CC3CC(C2)CC1C3
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
Percent Purity 
  • (3)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (3)
  • (2)
  • (2)