accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 105300-38-7

IUPAC Name: 6-fluoro-4H-chromen-4-one
Synonyms: 6-fluorochromen-4-one
Molecular Weight (g/mol): 164.14
Molecular Formula: C9H5FO2
InChi Key: WHIGSYZUTMYUAX-UHFFFAOYSA-N
SMILES: FC1=CC=C2OC=CC(=O)C2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results