accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH3NH3NH3NH3NH3NH3ClClClCo

CAS RN 10534-89-1

IUPAC Name: cobalt(3+) hexaamine trichloride
Synonyms: hexamminecobalt(III) chloride
Molecular Weight (g/mol): 267.47
Molecular Formula: Cl3CoH18N6
InChi Key: JXBGZYGSWFSYFI-UHFFFAOYSA-K
SMILES: N.N.N.N.N.N.[Cl-].[Cl-].[Cl-].[Co+3]
Quantity 
  • (1)
  • (3)
  • (3)
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (6)
Physical Form 
  • (2)
  • (5)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (5)
  • (1)
  • (1)
  • (3)
  • (3)
  • (2)
  • (1)
  • (2)
  • (6)
  • (2)
  • (5)
  • (2)