accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 105365-51-3

H3COBOHOH

CAS RN 105365-51-3

IUPAC Name: (4-butoxyphenyl)boronic acid
Synonyms: 4-butoxyphenylboronic acid
Molecular Weight (g/mol): 194.04
Molecular Formula: C10H15BO3
InChi Key: QUPFQMXWFNJUNJ-UHFFFAOYSA-N
SMILES: CCCCOC1=CC=C(C=C1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results