accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 1053655-59-6

ONNH2N

CAS RN 1053655-59-6

IUPAC Name: 1-(oxan-2-yl)-1H-indazol-6-amine
Synonyms: 1-(oxan-2-yl)indazol-6-amine
Molecular Weight (g/mol): 217.27
Molecular Formula: C12H15N3O
InChi Key: BVMGAUOJVUEFSV-UHFFFAOYNA-N
SMILES: NC1=CC=C2C=NN(C3CCCCO3)C2=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results