accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 10543-42-7

OOSOOCl

CAS RN 10543-42-7

IUPAC Name: 2-oxo-2H-chromene-6-sulfonyl chloride
Synonyms: 2-oxochromene-6-sulfonyl chloride
Molecular Weight (g/mol): 244.65
Molecular Formula: C9H5ClO4S
InChi Key: HQIPMBGUDSOVEA-UHFFFAOYSA-N
SMILES: ClS(=O)(=O)C1=CC=C2OC(=O)C=CC2=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results