accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CCH3BrNH2Br

CAS RN 10546-65-3

IUPAC Name: 2,6-dibromo-4-(propan-2-yl)aniline
Synonyms: 2,6-dibromo-4-isopropylaniline
Molecular Weight (g/mol): 293
Molecular Formula: C9H11Br2N
InChi Key: CJEBZUFROMNDEK-UHFFFAOYSA-N
SMILES: CC(C)C1=CC(Br)=C(N)C(Br)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results