accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 105494-65-3

CH3SnCH3H3CH3C(E/Z)

CAS RN 105494-65-3

IUPAC Name: tributyl(prop-1-en-1-yl)stannane
Synonyms: tributyl(prop-1-en-1-yl)stannane
Molecular Weight (g/mol): 331.13
Molecular Formula: C15H32Sn
InChi Key: FJXYMVBFBYAWDR-UHFFFAOYSA-N
SMILES: CCCC[Sn](CCCC)(CCCC)C=CC
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)