accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 105536-22-9

H3CCH3H3CCH3SOONH2CH3CH3

CAS RN 105536-22-9

IUPAC Name: 2,4,6-tris(propan-2-yl)benzene-1-sulfonamide
Synonyms: 2,4,6-triisopropylbenzenesulfonamide
Molecular Weight (g/mol): 283.43
Molecular Formula: C15H25NO2S
InChi Key: PRMNQLMPSVOZIX-UHFFFAOYSA-N
SMILES: CC(C)C1=CC(C(C)C)=C(C(=C1)C(C)C)S(N)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results