accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3ONH3CBr

CAS RN 10558-25-5

IUPAC Name: 4-bromo-3,5-dimethyl-1,2-oxazole
Synonyms: 4-bromo-3,5-dimethyl-1,2-oxazole
Molecular Weight (g/mol): 176.01
Molecular Formula: C5H6BrNO
InChi Key: GYHZPSUAMYIFQD-UHFFFAOYSA-N
SMILES: CC1=C(Br)C(C)=NO1
Quantity 
  • (2)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results