accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CCH3H3CPSOOHOCH3CH3H3C

CAS RN 1055888-89-5

IUPAC Name: 3-(di-tert-butylphosphanyl)propane-1-sulfonic acid
Synonyms: 3-(di-tert-butylphosphanyl)propane-1-sulfonic acid
Molecular Weight (g/mol): 268.35
Molecular Formula: C11H25O3PS
InChi Key: JPNPRWMRUCIEMN-UHFFFAOYSA-N
SMILES: CC(C)(C)P(CCCS(O)(=O)=O)C(C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results