accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CONOO

CAS RN 105596-63-2

IUPAC Name: 2-methoxypyridine-4-carboxylate
Synonyms: 2-methoxypyridine-4-carboxylate
Molecular Weight (g/mol): 152.13
Molecular Formula: C7H6NO3
InChi Key: DPNDWFVORIGXQO-UHFFFAOYSA-M
SMILES: COC1=CC(=CC=N1)C([O-])=O
Percent Purity 
  • (2)
  • (6)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (6)
  • (2)
  • (6)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)